CAS RN 38350-87-7
IUPAC Name:
4-heptylbenzoic acid
Synonyms:
4-heptylbenzoic acid
Molecular Weight (g/mol):
220.31
Molecular Formula:
C14H20O2
InChi Key:
VSUKEWPHURLYTK-UHFFFAOYSA-N
SMILES:
CCCCCCCC1=CC=C(C=C1)C(O)=O
1
–
1
of
1
results
1
4-n-Heptylbenzoic acid, 99+%
CAS: 38350-87-7 Molecular Formula: C14H20O2 Molecular Weight (g/mol): 220.312 MDL Number: MFCD00009722 InChI Key: VSUKEWPHURLYTK-UHFFFAOYSA-N Synonym: benzoic acid, 4-heptyl,4-n-heptylbenzoic acid,4-heptylbenzoicacid,4-heptylbenzoate,p-heptylbenzoic acid,acmc-209izm,4-n-heptyl-benzoic acid,4-heptylbenzoic acid PubChem CID: 170036 IUPAC Name: 4-heptylbenzoic acid SMILES: CCCCCCCC1=CC=C(C=C1)C(=O)O