accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 384-22-5

IUPAC Name: 1-nitro-2-(trifluoromethyl)benzene
Synonyms: 1-NO2 2-CF3 benzene
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: NDZJSUCUYPZXPR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results