accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 38462-22-5

CH3OCH3H3CSH

CAS RN 38462-22-5

IUPAC Name: 5-methyl-2-(2-sulfanylpropan-2-yl)cyclohexan-1-one
Synonyms: 5-methyl-2-(2-sulfanylpropan-2-yl)cyclohexan-1-one
Molecular Weight (g/mol): 186.31
Molecular Formula: C10H18OS
InChi Key: RVOKNSFEAOYULQ-UHFFFAOYNA-N
SMILES: CC1CCC(C(=O)C1)C(C)(C)S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results