accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 38465-86-0

IUPAC Name: cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide bis(methyldiphenylphosphane) iridium
Synonyms: 1,5-cyclooctadiene bis(methyldiphenylphosphane) hexafluorophosphate iridium
Molecular Weight (g/mol): 845.81
Molecular Formula: C34H38F6IrP3
InChi Key: MUHONFFTSOCMDH-UHFFFAOYSA-N
SMILES: [Ir].F[P-](F)(F)(F)(F)F.C1CC=CCCC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)