accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ClCH3ClH3CCl

CAS RN 3849-01-2

IUPAC Name: 1,3,5-tris(chloromethyl)-2,4,6-trimethylbenzene
Synonyms: 1,3,5-tris(chloromethyl)-2,4,6-trimethylbenzene
Molecular Weight (g/mol): 265.6
Molecular Formula: C12H15Cl3
InChi Key: PHQFMPNZCIHSPC-UHFFFAOYSA-N
SMILES: CC1=C(CCl)C(C)=C(CCl)C(C)=C1CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results