accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NONOO

CAS RN 38560-30-4

IUPAC Name: 1-(4-nitrophenyl)piperidin-2-one
Synonyms: 1-(4-nitrophenyl)piperidin-2-one
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: VPCQXIGQMFWXII-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1CCCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results