CAS RN 385786-48-1
CAS RN 385786-48-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Adaptaquin
HIF-prolyl hydroxylase-2 (PHD2) inhibitor; antioxidant