CAS RN 38597-28-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-ribo Phytosphingosine 1-Phosphate, TRC
CAS: 38597-28-3 Molecular Formula: C18 H40 N O6 P Molecular Weight (g/mol): 397.49 Synonym: (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol 1-(Dihydrogen Phosphate); IUPAC Name: [(2S,3S,4R)-2-amino-3,4-dihydroxyoctadecyl] dihydrogen phosphate SMILES: CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@@H](N)COP(=O)(O)O