accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 38614-36-7

IUPAC Name: magnesium(2+) 2-methylprop-1-en-1-ide bromide
Synonyms: magnesium(2+) 2-methylprop-1-en-1-ide bromide
Molecular Weight (g/mol): 159.31
Molecular Formula: C4H7BrMg
InChi Key: FFUIUNZKXBJVSC-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC(C)=[CH-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results