accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 387-44-0

IUPAC Name: 7-fluoro-1H-indole
Synonyms: 7-fluoro-1H-indole
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: XONKJZDHGCMRRF-UHFFFAOYSA-N
SMILES: FC1=C2NC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results