accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFHSCu

CAS RN 3872-23-9

IUPAC Name: trifluoromethanethiol copper
Synonyms: trifluoromethanethiol copper
Molecular Weight (g/mol): 165.62
Molecular Formula: CHCuF3S
InChi Key: JHTKDYFNOCMNDU-UHFFFAOYSA-N
SMILES: [Cu].FC(F)(F)S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)