CAS RN 38748-32-2
CAS RN 38748-32-2
IUPAC Name:
(1S,2S,4S,5S,7R,8R,9S,11S,13S)-8-hydroxy-1-methyl-7-(propan-2-yl)-3,6,10,16-tetraoxaheptacyclo[11.7.0.0²,⁴.0²,⁹.0⁵,⁷.0⁹,¹¹.0¹⁴,¹⁸]icos-14(18)-en-17-one
Synonyms:
triptolide triptolid triptolide (substance) triptolide (9CI)
Molecular Weight (g/mol):
360.41
Molecular Formula:
C20H24O6
InChi Key:
DFBIRQPKNDILPW-CIVMWXNOSA-N
SMILES:
CC(C)[C@]12O[C@H]1[C@@H]1O[C@]11[C@]3(O[C@H]3C[C@H]3C4=C(CC[C@]13C)C(=O)OC4)[C@@H]2O
1
–
2
of
2
results
1
Thermo Scientific Chemicals Triptolide
CAS: 38748-32-2 Molecular Formula: C20H24O6
2
Tocris Bioscience™ Triptolide
CAS: 38748-32-2 Molecular Formula: C20H24O6