accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3886-70-2

IUPAC Name: (1R)-1-(naphthalen-1-yl)ethan-1-aminium
Synonyms: (1R)-1-(naphthalen-1-yl)ethanaminium
Molecular Weight (g/mol): 172.25
Molecular Formula: C12H14N
InChi Key: RTCUCQWIICFPOD-SECBINFHSA-O
SMILES: C[C@@H]([NH3+])C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (8)
Quantity 
  • (4)
  • (3)
  • (5)
Boiling Point 
  • (8)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (8)
  • (4)
  • (3)
  • (5)
  • (8)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)