accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOHOOCH3N

CAS RN 389-08-2

IUPAC Name: 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Synonyms: nalidixic acid dixinal negram NegGram nalidixic acid (substance) nalurin urodixin acide nalidixico nalidixic acid USP27 dixiben
Molecular Weight (g/mol): 232.24
Molecular Formula: C12H12N2O3
InChi Key: MHWLWQUZZRMNGJ-UHFFFAOYSA-N
SMILES: CCN1C=C(C(O)=O)C(=O)C2=CC=C(C)N=C12
Molecular Weight (g/mol) 
  • (6)
  • (9)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (1)
  • (5)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (6)
  • (9)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)