accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2OH2C

CAS RN 38945-21-0

IUPAC Name: hydrogen O-(prop-2-en-1-yl)hydroxylamine chloride
Synonyms: hydrogen O-(prop-2-en-1-yl)hydroxylamine chloride
Molecular Weight (g/mol): 109.55
Molecular Formula: C3H8ClNO
InChi Key: XIQUJVRFXPBMHS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NOCC=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)