accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 389621-81-2

BONHOOH

CAS RN 389621-81-2

IUPAC Name: [4-(pyrrolidine-1-carbonyl)phenyl]boronic acid
Synonyms: 4-(pyrrolidine-1-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 219.05
Molecular Formula: C11H14BNO3
InChi Key: VKPBESPVHGDDJS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results