accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3902-71-4

IUPAC Name: 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
Synonyms: trioxsalen trioxisaleno 2',4,8-trimethylpsoralen lactone trioxsalen (substance) 4,5',8-trimethylpsoralene 4,8,5'-trimethylpsoralen trioxysalen trisoralen benzofuran derivative
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: FMHHVULEAZTJMA-UHFFFAOYSA-N
SMILES: CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)