accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39067-29-3

IUPAC Name: 2-(pyridin-3-yl)-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(pyridin-3-yl)-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 206.22
Molecular Formula: C9H6N2O2S
InChi Key: FOQFGMAZUTUELM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC(=N1)C1=CN=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)