accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39082-23-0

TeHfTe

CAS RN 39082-23-0

IUPAC Name: ditellanylidenehafnium
Synonyms: ditellanylidenehafnium
Molecular Weight (g/mol): 433.69
Molecular Formula: HfTe2
InChi Key: BCMXGUYIVFZMCT-UHFFFAOYSA-N
SMILES: [Te]=[Hf]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results