accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSONH

CAS RN 39093-93-1

IUPAC Name: 1λ⁶-thiomorpholine-1,1-dione
Synonyms: 1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 135.18
Molecular Formula: C4H9NO2S
InChi Key: NDOVLWQBFFJETK-UHFFFAOYSA-N
SMILES: O=S1(=O)CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results