accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3N

CAS RN 39101-54-7

IUPAC Name: 2-(3,5-dimethylphenyl)acetonitrile
Synonyms: 2-(3,5-dimethylphenyl)acetonitrile
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: LMUKNQSVBFEUKR-UHFFFAOYSA-N
SMILES: CC1=CC(CC#N)=CC(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)