CAS RN 3914-34-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isouramil, TRC
CAS: 3914-34-9 Molecular Formula: C4H5N3O3 Molecular Weight (g/mol): 143.1 Synonym: 5,6-Dihydroxycytosine,6-Amino-5-hydroxy-2,4(1H,3H)-pyrimidinedione,6-Aminoisobarbituric Acid; IUPAC Name: 6-amino-5-hydroxy-1H-pyrimidine-2,4-dione SMILES: NC1=C(O)C(=O)NC(=O)N1