accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39148-24-8

IUPAC Name: aluminium(3+) tris((phosphooxy)ethane)
Synonyms: fosetyl-al
Molecular Weight (g/mol): 351.08
Molecular Formula: C6H15AlO9P3
InChi Key: OIPMQULDKWSNGX-UHFFFAOYSA-N
SMILES: [Al+3].CCOP(=O)=O.CCOP(=O)=O.CCOP(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results