accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNCH3

CAS RN 39180-84-2

IUPAC Name: methyl[(3-methylphenyl)methyl]amine
Synonyms: methyl[(3-methylphenyl)methyl]amine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: WCTNVGNEUDTSOZ-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results