accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHHNO

CAS RN 3922-40-5

IUPAC Name: 1,4,7,10-tetrahydro-1,10-phenanthroline-4,7-dione
Synonyms: 1,10-dihydro-1,10-phenanthroline-4,7-dione
Molecular Weight (g/mol): 212.21
Molecular Formula: C12H8N2O2
InChi Key: SLIBCJURSADKPV-UHFFFAOYSA-N
SMILES: O=C1C=CNC2=C3NC=CC(=O)C3=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results