accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39222-73-6

CH3H3CCH3SNH2NN

CAS RN 39222-73-6

IUPAC Name: 5-tert-butyl-1,3,4-thiadiazol-2-amine
Synonyms: 5-tert-butyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 157.24
Molecular Formula: C6H11N3S
InChi Key: ICXDPEFCLDSXLI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=NN=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results