accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3NClCl

CAS RN 39226-95-4

IUPAC Name: (2,3-dichlorophenyl)methanaminium
Synonyms: (2,3-dichlorophenyl)methanaminium
Molecular Weight (g/mol): 177.05
Molecular Formula: C7H8Cl2N
InChi Key: JHBVZGONNIVXFJ-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=CC(Cl)=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results