accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39244-08-1

IUPAC Name: 3-chlorothieno[2,3-b]thiophene-2-carboxylate
Synonyms: 3-chlorothieno[2,3-b]thiophene-2-carboxylate
Molecular Weight (g/mol): 217.66
Molecular Formula: C7H2ClO2S2
InChi Key: GCQFYYGUNYJMNY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(Cl)C2=C(SC=C2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results