accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3937-96-0

IUPAC Name: 2-(4-methylbenzenesulfonyl)acetic acid
Synonyms: (4-methylbenzenesulfonyl)acetic acid
Molecular Weight (g/mol): 214.24
Molecular Formula: C9H10O4S
InChi Key: AQDHXMBUTDLAMD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results