accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OORhORhO

CAS RN 39373-27-8

IUPAC Name: oxo[(oxorhodio)oxy]rhodium pentahydrate
Synonyms: rhodium-platinum oxide
Molecular Weight (g/mol): 343.88
Molecular Formula: H10O8Rh2
InChi Key: XZIIPNYQAMUKBU-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O=[Rh]O[Rh]=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results