accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHSOOCH2H2CCH2

CAS RN 39389-20-3

IUPAC Name: 1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
Synonyms: 2-ethenylbenzenesulfonic acid; divinylbenzene
Molecular Weight (g/mol): 314.4
Molecular Formula: C18H18O3S
InChi Key: SIWVGXQOXWGJCI-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)