accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 394-31-0

IUPAC Name: 2-amino-5-hydroxybenzoic acid
Synonyms: 2-amino-5-hydroxybenzoic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: HYNQTSZBTIOFKH-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results