accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFNOO

CAS RN 394-41-2

IUPAC Name: 3-fluoro-4-nitrobenzen-1-olate
Synonyms: 3-fluoro-4-nitrobenzenolate
Molecular Weight (g/mol): 156.09
Molecular Formula: C6H3FNO3
InChi Key: CSSGKHVRDGATJL-UHFFFAOYSA-M
SMILES: [O-]C1=CC=C(C(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results