accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOOOOOOOOHOHMgMgMgMgMgH2OH2OH2OH2O

CAS RN 39409-82-0

IUPAC Name: magnesium(2+) hydroxide carbonate
Synonyms: magnesium carbonate hydroxide
Molecular Weight (g/mol): 101.32
Molecular Formula: CHMgO4
InChi Key: OUHCLAKJJGMPSW-UHFFFAOYSA-K
SMILES: [OH-].[Mg++].[O-]C([O-])=O
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (11)
Percent Purity 
  • (1)
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (11)
  • (1)
  • (3)
  • (2)
  • (3)