accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39416-48-3

BrBrBrHN

CAS RN 39416-48-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 319.82
Molecular Formula: C5H6Br3N
InChi Key: GWJBDYRGIFGTOG-UHFFFAOYSA-O
SMILES: [H+].[Br].[Br].[Br].C1=CC=NC=C1
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
Grade 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)