accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSOOO(R)(R)(R)HNH3COO(R)HOO(R)HO(S)HOO(S)O(R)HO(R)OHNaA

CAS RN 39455-18-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: (C14H20NO14S)ANa
InChi Key:
SMILES: [Na+].[H][C@@]1(O[C@@H]2[C@@H](NC(C)=O)C(-*)O[C@H](CO)[C@@H]2OS([O-])(=O)=O)O[C@@H]([C@@H](O-*)[C@H](O)[C@H]1O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results