accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39515-51-0

IUPAC Name: 3-phenoxybenzaldehyde
Synonyms: 3-phenoxybenzaldehyde
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: MRLGCTNJRREZHZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(OC2=CC=CC=C2)=C1
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)