accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCO

CAS RN 3954-13-0

IUPAC Name: 1-isocyanatopentane
Synonyms: 1-isocyanatopentane
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: VRVUKQWNRPNACD-UHFFFAOYSA-N
SMILES: CCCCCN=C=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results