accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39551-54-7

IUPAC Name: 2,4-dichloropyrido[3,2-d]pyrimidine
Synonyms: 2,4-dichloropyrido[3,2-d]pyrimidine
Molecular Weight (g/mol): 200.02
Molecular Formula: C7H3Cl2N3
InChi Key: UVVFNZJVLRJSMW-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=CN=C2C(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results