accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH

CAS RN 39593-08-3

IUPAC Name: 1-(4-methylphenyl)piperazine
Synonyms: 1-(4-methylphenyl)piperazine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: ONEYFZXGNFNRJH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N1CCNCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results