accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 396-30-5

IUPAC Name: 6-fluoroquinoline
Synonyms: 6-fluoroquinoline
Molecular Weight (g/mol): 147.15
Molecular Formula: C9H6FN
InChi Key: RMDCSDVIVXJELQ-UHFFFAOYSA-N
SMILES: FC1=CC=C2N=CC=CC2=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results