accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOHCl

CAS RN 3964-57-6

IUPAC Name: methyl 3-chloro-4-hydroxybenzoate
Synonyms: methyl 3-chloro-4-hydroxybenzoate
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: ZSBIMTDWIGWJPW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results