accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39648-67-4

OPOOO

CAS RN 39648-67-4

IUPAC Name: 13-oxo-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-olate
Synonyms: 13-oxo-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-olate
Molecular Weight (g/mol): 347.29
Molecular Formula: C20H12O4P
InChi Key: JEHUZVBIUCAMRZ-UHFFFAOYSA-M
SMILES: [O-]P1(=O)OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results