accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3966-32-3

CH3O(R)OO

CAS RN 3966-32-3

IUPAC Name: (2R)-2-methoxy-2-phenylacetate
Synonyms: (2R)-2-methoxy-2-phenylacetate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: DIWVBIXQCNRCFE-MRVPVSSYSA-M
SMILES: CO[C@@H](C([O-])=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)