accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3CH3H3COH2N

CAS RN 39684-28-1

IUPAC Name: hydrogen O-tert-butylhydroxylamine chloride
Synonyms: hydrogen O-tert-butylhydroxylamine chloride
Molecular Weight (g/mol): 125.6
Molecular Formula: C4H12ClNO
InChi Key: ZBDXGNXNXXPKJI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)ON
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)