accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(S)(R)(S)OO(R)OO

CAS RN 39746-00-4

IUPAC Name: (3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-hexahydro-2H-cyclopenta[b]furan-5-yl benzoate
Synonyms: (3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-hexahydrocyclopenta[b]furan-5-yl benzoate
Molecular Weight (g/mol): 276.29
Molecular Formula: C15H16O5
InChi Key: OBRRYUZUDKVCOO-FVCCEPFGSA-N
SMILES: OC[C@H]1[C@@H](C[C@@H]2OC(=O)C[C@H]12)OC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results