accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3976-69-0

IUPAC Name: methyl 3-hydroxybutanoate
Synonyms: methyl 3-hydroxybutyrate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: LDLDJEAVRNAEBW-UHFFFAOYNA-N
SMILES: COC(=O)CC(C)O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)