accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3977-29-5

IUPAC Name: 2-amino-6-methyl-1,4-dihydropyrimidin-4-one
Synonyms: 2-amino-6-methylpyrimidin-4-ol
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: KWXIPEYKZKIAKR-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N=C(N)N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)