accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3N

CAS RN 3978-81-2

IUPAC Name: 4-tert-butylpyridine
Synonyms: 4-(tert-butyl)pyridine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: YSHMQTRICHYLGF-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=NC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results