CAS RN 39785-37-0
IUPAC Name:
4-methoxy-3,5-dimethylaniline
Synonyms:
4-methoxy-3,5-dimethylaniline
Molecular Weight (g/mol):
151.21
Molecular Formula:
C9H13NO
InChi Key:
HUPGZEYAAFYLSC-UHFFFAOYSA-N
SMILES:
COC1=C(C)C=C(N)C=C1C
1
–
1
of
1
results
1
4-Methoxy-3,5-dimethylaniline, 98%
CAS: 39785-37-0 Molecular Formula: C9H13NO Molecular Weight (g/mol): 151.209 MDL Number: MFCD01907075 InChI Key: HUPGZEYAAFYLSC-UHFFFAOYSA-N Synonym: 4-amino-2,6-dimethylanisole,4-methoxy-3,5-dimethyl-benzenamine,3,5-dimethyl-4-methoxyaniline,4-methoxy-3,5-dimethylphenyl amine,4-methoxy-3,5-dimethyl-aniline,4-methoxy-3,5-dimethylphenylamine,benzenamine, 4-methoxy-3,5-dimethyl,4-methoxy-3,5-xylidine,2,6-dimethyl-p-anisidine,3,5-dimethyl-p-anisidine PubChem CID: 4166097 IUPAC Name: 4-methoxy-3,5-dimethylaniline SMILES: CC1=CC(=CC(=C1OC)C)N